siRNA Details
| Gene Symbol: Gm10639 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122660.1 | NP_001116132.1 | 5 | 398 | ||
| GenBank mRNA | BC030173.1 | - | 5 | 409 | 5041881 | |
| Gene Symbol: Gm3776 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243092.1 | NP_001230021.1 | 5 | 398 | ||
| GenBank mRNA | BC061134.1 | - | 368 | 30308304 | ||
| Gene Symbol: Gsta1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008181.3 | NP_032207.3 | 5 | 385 | ||
| GenBank mRNA | BC132572.1 | - | 368 | 40130849 | ||
| BC132576.1 | - | 368 | 40130853 | |||
| CJ228212.1 | - | 3 | 242 | |||
| Gene Symbol: Gsta2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008182.3 | NP_032208.2 | 5 | 380 | ||
| GenBank mRNA | AK002712.1 | - | 5 | 387 | ||
| BC061133.1 | - | 4 | 323 | 30303519 | ||
| J03958.1 | - | 5 | 395 | |||

Gm10639
Genomic Map