siRNA Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289502.1 | NP_001276431.1 | 5, 6 | 1097 | ||
| NM_001289503.1 | NP_001276432.1 | 6, 7 | 829 | |||
| NM_178629.6 | NP_848744.2 | 6, 7 | 1247 | |||
| GenBank mRNA | AK016317.1 | - | 6, 7 | 796 | ||
| AK045940.1 | - | 6, 7 | 1249 | |||
| AK051322.1 | - | 5, 6 | 792 | |||
| BC024061.1 | - | 5, 6 | 529 | |||
| BC050993.1 | - | 6, 7 | 1085 | |||
| BC086665.1 | - | 6, 7 | 1178 | 30612014 | ||

Ispd
Genomic Map