siRNA Details
| Gene Symbol: Ispd | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289502.1 | NP_001276431.1 | 9 | 1437 | ||
| NM_001289503.1 | NP_001276432.1 | 10 | 1169 | |||
| NM_178629.6 | NP_848744.2 | 10 | 1587 | |||
| GenBank mRNA | AK016317.1 | - | 10 | 1135 | ||
| AK051322.1 | - | 9 | 1132 | |||
| BC024061.1 | - | 9 | 869 | |||
| BC049756.1 | - | 322 | ||||
| BC050993.1 | - | 10 | 1425 | |||
| BC086665.1 | - | 10 | 1518 | 30612014 | ||

Ispd
Genomic Map