siRNA Details
| Gene Symbol: Gm13275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085533.2 | NP_001079002.1 | 1 | 1511 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Gm13275| Gene Symbol: Gm13275 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085533.2 | NP_001079002.1 | 1 | 1511 | ||