siRNA Details
| Gene Symbol: Zfp943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025373.2 | NP_001020544.1 | 4 | 758 | ||
| GenBank mRNA | BC066031.1 | - | 4 | 559 | 6418362 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp943| Gene Symbol: Zfp943 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001025373.2 | NP_001020544.1 | 4 | 758 | ||
| GenBank mRNA | BC066031.1 | - | 4 | 559 | 6418362 | |