siRNA Details
| Gene Symbol: Tox | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145711.4 | NP_663757.3 | 5, 6 | 1153 | ||
| GenBank mRNA | AF472514.1 | - | 5, 6 | 949 | ||
| AK029694.1 | - | 5, 6 | 950 | |||
| AK038671.1 | - | 5, 6 | 1156 | |||
| AK133397.1 | - | 5, 6 | 950 | |||
| AK161578.1 | - | 5, 6 | 939 | |||
| BC038873.1 | - | 1, 2 | 169 | |||
| BC080732.1 | - | 5, 6 | 1070 | 6518356 | ||

Tox
Genomic Map