siRNA Details
Gene Symbol: Cela3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126318.1 | NP_001119790.1 | 3, 4 | 258 | ||
GenBank mRNA | BM501883.1 | - | 198 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Cela3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001126318.1 | NP_001119790.1 | 3, 4 | 258 | ||
GenBank mRNA | BM501883.1 | - | 198 |