siRNA Details
| Gene Symbol: Sfmbt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198808.1 | NP_001185737.1 | 7 | 833 | ||
| NM_177386.5 | NP_796360.2 | 8 | 956 | |||
| GenBank mRNA | AK052339.1 | - | 7 | 899 | ||
| AK147557.1 | - | 7 | 838 | |||
| AK147651.1 | - | 8 | 811 | |||
| AK220408.1 | - | 5 | 698 | |||
| BC139354.1 | - | 7 | 744 | 8862376 | ||
| BC145610.1 | - | 6 | 700 | 9054176 | ||

Sfmbt2
Genomic Map