siRNA Details
Gene Symbol: Gm13051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 2578 | ||
GenBank mRNA | BC108336.1 | - | 4 | 2556 | 30462068 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm13051 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 2578 | ||
GenBank mRNA | BC108336.1 | - | 4 | 2556 | 30462068 |