siRNA Details
| Gene Symbol: Gm13051 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037926.2 | NP_001033015.1 | 4 | 2679 | ||
| GenBank mRNA | BC108336.1 | - | 4 | 2657 | 30462068 | |
| Gene Symbol: Gm13152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC066875.1 | - | 2519 | 30111586 | ||

Gm13051
Genomic Map