siRNA Details
| Gene Symbol: Myo5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081322.1 | NP_001074791.1 | 33 | 4104 | ||
| GenBank mRNA | AK033571.1 | - | 9 | 909 | ||
| BC003985.1 | - | 4 | 560 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo5c| Gene Symbol: Myo5c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081322.1 | NP_001074791.1 | 33 | 4104 | ||
| GenBank mRNA | AK033571.1 | - | 9 | 909 | ||
| BC003985.1 | - | 4 | 560 | |||