siRNA Details
| Gene Symbol: Sbk1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145587.2 | NP_663562.1 | 2, 3 | 712 | ||
| GenBank mRNA | BC024114.1 | - | 2, 3 | 453 | 5098034 | |
| BC025837.1 | - | 2, 3 | 549 | 5135723 | ||
| BC031759.1 | - | 2, 3 | 712 | 4194152 | ||

Sbk1
Genomic Map