siRNA Details
| Gene Symbol: Cyp26a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_007811.2 | NP_031837.2 | 6, 7 | 1254 | ||
| GenBank mRNA | AA239785.1 | - | 2, 3 | 183 | ||
| AF115769.1 | - | 6, 7 | 1178 | |||
| BC012673.1 | - | 6, 7 | 1214 | 4210893 | ||
| Y12657.1 | - | 6, 7 | 1208 | |||

Cyp26a1
Genomic Map