siRNA Details
Gene Symbol: Gm5431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024230.2 | NP_001019401.2 | 5 | 1137 | ||
GenBank mRNA | AK144287.1 | - | 5 | 1138 | ||
AK165745.1 | - | 5 | 1138 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Gm5431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024230.2 | NP_001019401.2 | 5 | 1137 | ||
GenBank mRNA | AK144287.1 | - | 5 | 1138 | ||
AK165745.1 | - | 5 | 1138 |