siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 28 | 5537 | ||
| GenBank mRNA | AK051474.1 | - | 5 | 765 | ||
| AK122351.1 | - | 25 | 4527 | |||
| AK147675.1 | - | 26 | 6127 | |||
| AK162968.1 | - | 7 | 997 | |||

Cul9
Genomic Map