siRNA Details
| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 36 | 7054 | ||
| GenBank mRNA | AK051474.1 | - | 13 | 2282 | ||
| AK122351.1 | - | 33 | 6044 | |||
| AK162968.1 | - | 15 | 2515 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cul9| Gene Symbol: Cul9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081335.2 | NP_001074804.2 | 36 | 7054 | ||
| GenBank mRNA | AK051474.1 | - | 13 | 2282 | ||
| AK122351.1 | - | 33 | 6044 | |||
| AK162968.1 | - | 15 | 2515 | |||