siRNA Details
| Gene Symbol: D130043K22Rik | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AK051381.1 | - | 5 | 1322 | ||
| AK122246.1 | - | 5 | 1319 | |||
| AK148668.1 | - | 3 | 926 | |||
| BC115723.1 | - | 5 | 1238 | 40086618 | ||
| BC115940.1 | - | 5 | 1238 | 40086616 | ||

D130043K22Rik
Genomic Map