siRNA Details
| Gene Symbol: Eif4a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001123037.2 | NP_001116509.1 | 6 | 921 | ||
| NM_001123038.2 | NP_001116510.1 | 7 | 1061 | |||
| NM_013506.3 | NP_038534.2 | 7 | 1061 | |||
| GenBank mRNA | AK076509.1 | - | 7 | 772 | ||
| AK088669.1 | - | 7 | 773 | |||
| AK132969.1 | - | 7 | 772 | |||
| AK165898.1 | - | 6 | 632 | |||
| BC094422.1 | - | 7 | 757 | 6808404 | ||
| CK623185.1 | - | 2 | 168 | |||
| X12507.1 | - | 7 | 773 | |||

Eif4a2
Genomic Map