siRNA Details
Gene Symbol: Dip2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081419.2 | NP_001074888.2 | 3 | 282 | ||
GenBank mRNA | AK052899.1 | - | 3 | 224 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Dip2a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081419.2 | NP_001074888.2 | 3 | 282 | ||
GenBank mRNA | AK052899.1 | - | 3 | 224 |