siRNA Details
| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080706.1 | NP_001074175.1 | 28 | 4256 | ||
| GenBank mRNA | AB287295.1 | - | 28 | 3981 | ||
| AK088840.1 | - | 3 | 185 | |||
| BC094345.1 | - | 9 | 1373 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Btaf1| Gene Symbol: Btaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001080706.1 | NP_001074175.1 | 28 | 4256 | ||
| GenBank mRNA | AB287295.1 | - | 28 | 3981 | ||
| AK088840.1 | - | 3 | 185 | |||
| BC094345.1 | - | 9 | 1373 | |||