siRNA Details
Gene Symbol: Zfp932 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_145563.2 | NP_663538.2 | 4 | 1536 | ||
GenBank mRNA | AK032220.1 | - | 4 | 1077 | ||
BC012405.1 | - | 4 | 1522 |
Gene Symbol: Gm20939 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001024731.2 | NP_001019902.2 | 5 | 1356 | ||
GenBank mRNA | BC070434.1 | - | 5 | 1356 | 6851340 |
