siRNA Details
| Gene Symbol: Ttc27 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_152817.4 | NP_690030.3 | 13 | 1671 | ||
| GenBank mRNA | AK012131.1 | - | 13 | 1671 | ||
| AK030942.1 | - | 13 | 1672 | |||
| AK049324.1 | - | 13 | 1672 | |||
| AK168138.1 | - | 13 | 1662 | |||
| BC021912.1 | - | 12 | 1406 | 4503335 | ||

Ttc27
Genomic Map