siRNA Details
| Gene Symbol: Sptan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177667.1 | NP_001171138.1 | 16 | 2274 | ||
| NM_001177668.1 | NP_001171139.1 | 16 | 2274 | |||
| GenBank mRNA | AK132224.1 | - | 16 | 2158 | ||
| AK141149.1 | - | 16 | 2155 | |||
| AK147317.1 | - | 16 | 2158 | |||
| BC150941.1 | - | 16 | 2110 | 9087852 | ||
| BC158015.1 | - | 16 | 2110 | 9007321 | ||
| BC172095.1 | - | 16 | 2110 | 9054381 | ||

Sptan1
Genomic Map