siRNA Details
| Gene Symbol: Sptan1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177667.1 | NP_001171138.1 | 29 | 3937 | ||
| NM_001177668.1 | NP_001171139.1 | 28 | 3877 | |||
| GenBank mRNA | AK147317.1 | - | 28 | 3762 | ||
| AK164891.1 | - | 6 | 358 | |||
| BC027791.1 | - | 6 | 369 | |||
| BC150941.1 | - | 29 | 3773 | 9087852 | ||
| BC158015.1 | - | 29 | 3773 | 9007321 | ||
| BC172095.1 | - | 28 | 3713 | 9054381 | ||
| X12801.1 | - | 8 | 667 | |||

Sptan1
Genomic Map