siRNA Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081409.1 | NP_001074878.1 | 4 | 639 | ||
| GenBank mRNA | AK032638.1 | - | 4 | 642 | ||
| AK047044.1 | - | 4 | 652 | |||
| BC052212.1 | - | 4 | 640 | |||
| BC061492.1 | - | 4 | 583 | |||
| BC082777.1 | - | 4 | 631 | |||
| BC145689.1 | - | 3 | 370 | 40130992 | ||

Phf20l1
Genomic Map