siRNA Details
| Gene Symbol: Phf20l1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001081409.1 | NP_001074878.1 | 11 | 1539 | ||
| GenBank mRNA | AK032638.1 | - | 11 | 1554 | ||
| AK040888.1 | - | 2 | 270 | |||
| AK041825.1 | - | 1 | 634 | |||
| AK080555.1 | - | 1 | 1171 | |||
| AK206721.1 | - | 2 | 230 | |||
| BC052212.1 | - | 10 | 1465 | |||
| BC061492.1 | - | 10 | 1405 | |||
| BC082777.1 | - | 11 | 1534 | |||
| BC145689.1 | - | 10 | 1270 | 40130992 | ||

Phf20l1
Genomic Map