siRNA Details
| Gene Symbol: Prg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110146.1 | NP_001103616.1 | 7 | 570 | ||
| NM_021400.3 | NP_067375.3 | 8 | 2964 | |||
| GenBank mRNA | AB034730.1 | - | 9 | 2463 | ||
| AK132597.1 | - | 2 | 710 | |||
| AK149469.1 | - | 7 | 571 | |||
| BC130021.1 | - | 6 | 534 | |||

Prg4
Genomic Map