siRNA Details
| Gene Symbol: Prg4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110146.1 | NP_001103616.1 | 2, 3 | 111 | ||
| NM_021400.3 | NP_067375.3 | 2, 3 | 111 | |||
| GenBank mRNA | AB034730.1 | - | 2, 3 | 111 | ||
| AK149469.1 | - | 2, 3 | 112 | |||

Prg4
Genomic Map