siRNA Details
| Gene Symbol: Xaf1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037713.4 | NP_001032802.2 | 5 | 659 | ||
| NM_001291153.1 | NP_001278082.1 | 3 | 374 | |||
| GenBank mRNA | BY726623.1 | - | 3 | 374 | ||
| CK130151.1 | - | 4 | 594 | |||
| EL957551.1 | - | 3 | 364 | |||

Xaf1
Genomic Map