siRNA Details
| Gene Symbol: Hist1h3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101954.1 | - | 1 | 963 | 40044770 | |
| BC101955.1 | - | 1 | 963 | 40044769 | ||
| BC101956.1 | - | 1 | 963 | 40044765 | ||
| BC103549.1 | - | 1 | 963 | 40044771 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h3f| Gene Symbol: Hist1h3f | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC101954.1 | - | 1 | 963 | 40044770 | |
| BC101955.1 | - | 1 | 963 | 40044769 | ||
| BC101956.1 | - | 1 | 963 | 40044765 | ||
| BC103549.1 | - | 1 | 963 | 40044771 | ||