siRNA Details
| Gene Symbol: Hacd3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021345.2 | NP_067320.2 | 10 | 1170 | ||
| GenBank mRNA | AK050556.1 | - | 10 | 1164 | ||
| AK051735.1 | - | 10 | 1129 | |||
| AK052865.1 | - | 10 | 1167 | |||
| AK159494.1 | - | 10 | 1136 | |||
| BC031755.1 | - | 10 | 1154 | 4488391 | ||
| BC057023.1 | - | 10 | 1146 | 6813640 | ||

Hacd3
Genomic Map