siRNA Details
| Gene Symbol: Srp54b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100109.1 | NP_001093579.1 | 11 | 1456 | ||
| GenBank mRNA | AK136033.1 | - | 11 | 1457 | ||
| X16319.1 | - | 1185 | ||||
| Gene Symbol: Srp54a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011899.4 | NP_036029.2 | 11 | 1220 | ||
| GenBank mRNA | AK011928.1 | - | 11 | 1184 | ||
| BC019683.1 | - | 11 | 1149 | 3708250 | ||

Srp54b
Genomic Map