siRNA Details
| Gene Symbol: Cd72 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110320.1 | NP_001103790.1 | 8 | 1078 | ||
| NM_001110321.1 | NP_001103791.1 | 7 | 1006 | |||
| NM_001110322.1 | NP_001103792.1 | 6 | 895 | |||
| NM_007654.2 | NP_031680.2 | 8 | 1057 | |||
| GenBank mRNA | AA203771.1 | - | 3 | 231 | ||
| AF543214.1 | - | 6 | 754 | |||
| AK134132.1 | - | 8 | 1081 | |||
| AK144191.1 | - | 7 | 1009 | |||
| AK151094.1 | - | 897 | ||||

Cd72
Genomic Map