siRNA Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183281.2 | NP_899104.2 | 3 | 381 | ||
| GenBank mRNA | AK036849.1 | - | 3 | 408 | ||
| BC150914.1 | - | 2 | 534 | |||
| BC150924.1 | - | 2 | 534 | |||
| BC172084.1 | - | 2 | 534 | 9054370 | ||

Tmem45a2
Genomic Map