siRNA Details
| Gene Symbol: Tmem45a2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_183281.2 | NP_899104.2 | 4 | 449 | ||
| GenBank mRNA | AK036849.1 | - | 4 | 476 | ||
| BC150914.1 | - | 3 | 602 | |||
| BC150924.1 | - | 3 | 602 | |||
| BC172084.1 | - | 3 | 602 | 9054370 | ||

Tmem45a2
Genomic Map