siRNA Details
| Gene Symbol: Gm10488 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099325.2 | NP_001092795.1 | 3 | 266 | ||
| GenBank mRNA | BC099541.1 | - | 3 | 266 | 30916030 | |
| Gene Symbol: Gm2030 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100445.2 | NP_001093915.1 | 3 | 297 | ||
| GenBank mRNA | AA061388.1 | - | 3 | 192 | ||

Gm10488
Genomic Map