siRNA Details
| Gene Symbol: Vmn1r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053219.2 | NP_444449.2 | 1 | 491 | ||
| GenBank mRNA | BC111822.1 | - | 1 | 490 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Vmn1r47| Gene Symbol: Vmn1r47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_053219.2 | NP_444449.2 | 1 | 491 | ||
| GenBank mRNA | BC111822.1 | - | 1 | 490 | ||