siRNA Details
| Gene Symbol: Hist1h2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175660.3 | NP_783591.2 | 1 | 449 | ||
| GenBank mRNA | BC117110.1 | - | 1 | 448 | 40126429 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Hist1h2ab| Gene Symbol: Hist1h2ab | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_175660.3 | NP_783591.2 | 1 | 449 | ||
| GenBank mRNA | BC117110.1 | - | 1 | 448 | 40126429 | |