siRNA Details
| Gene Symbol: Fam208b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_134063.3 | NP_598824.3 | 10 | 1510 | ||
| GenBank mRNA | AK138119.1 | - | 10 | 1511 | ||
| AK220437.1 | - | 10 | 1498 | |||
| BC075685.1 | - | 8 | 986 | |||
| BC085160.1 | - | 1 | 253 | |||

Fam208b
Genomic Map