siRNA Details
| Gene Symbol: LOC100045026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039241.2 | NP_001034330.1 | 5 | 1247 | ||
| GenBank mRNA | BC046609.1 | - | 5 | 1249 | 6397872 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC100045026| Gene Symbol: LOC100045026 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001039241.2 | NP_001034330.1 | 5 | 1247 | ||
| GenBank mRNA | BC046609.1 | - | 5 | 1249 | 6397872 | |