siRNA Details
| Gene Symbol: Zfp334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178411.3 | NP_848498.2 | 5 | 896 | ||
| GenBank mRNA | BC027792.1 | - | 1 | 231 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp334| Gene Symbol: Zfp334 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_178411.3 | NP_848498.2 | 5 | 896 | ||
| GenBank mRNA | BC027792.1 | - | 1 | 231 | ||