siRNA Details
Gene Symbol: Zfp334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178411.3 | NP_848498.2 | 5 | 940 | ||
GenBank mRNA | BC027792.1 | - | 1 | 275 | ||
BC040388.1 | - | 5 | 940 | 3501307 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Zfp334 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_178411.3 | NP_848498.2 | 5 | 940 | ||
GenBank mRNA | BC027792.1 | - | 1 | 275 | ||
BC040388.1 | - | 5 | 940 | 3501307 |