siRNA Details
| Gene Symbol: Slc7a5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011404.3 | NP_035534.2 | 3 | 784 | ||
| GenBank mRNA | AB017189.1 | - | 3 | 730 | ||
| AB023409.1 | - | 3 | 712 | |||
| AK142288.1 | - | 3 | 785 | |||
| AK148134.1 | - | 3 | 454 | |||
| AK180682.1 | - | 2 | 100 | |||
| AK183612.1 | - | 3 | 145 | |||
| AK199678.1 | - | 2 | 100 | |||
| AK203738.1 | - | 2 | 100 | |||
| BC026131.1 | - | 3 | 793 | 4188854 | ||

Slc7a5
Genomic Map