siRNA Details
| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 2 | 327 | ||
| GenBank mRNA | AK005247.1 | - | 2 | 329 | ||
| AK014081.1 | - | 2 | 277 | |||
| AK133585.1 | - | 2 | 325 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zfp593| Gene Symbol: Zfp593 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024215.2 | NP_077177.2 | 2 | 327 | ||
| GenBank mRNA | AK005247.1 | - | 2 | 329 | ||
| AK014081.1 | - | 2 | 277 | |||
| AK133585.1 | - | 2 | 325 | |||