siRNA Details
| Gene Symbol: Olfr725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146317.2 | NP_666429.2 | 1 | 757 | ||
| GenBank mRNA | BC125411.1 | - | 1 | 757 | 40129926 | |
| BC125417.1 | - | 1 | 757 | 40129932 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr725| Gene Symbol: Olfr725 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146317.2 | NP_666429.2 | 1 | 757 | ||
| GenBank mRNA | BC125411.1 | - | 1 | 757 | 40129926 | |
| BC125417.1 | - | 1 | 757 | 40129932 | ||