siRNA Details
| Gene Symbol: Rpl32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_172086.2 | NP_742083.1 | 2 | 129 | ||
| GenBank mRNA | AK002353.1 | - | 2 | 130 | ||
| AK011017.1 | - | 2 | 130 | |||
| AK012525.1 | - | 2 | 130 | |||
| AK028204.1 | - | 2 | 130 | |||
| AK146768.1 | - | 2 | 130 | |||
| AK148453.1 | - | 2 | 130 | |||
| AK150705.1 | - | 2 | 130 | |||
| AK168282.1 | - | 2 | 133 | |||
| BC030176.1 | - | 129 | ||||
| BC046339.1 | - | 2 | 98 | 6478080 | ||
| BC086909.1 | - | 2 | 133 | 30122780 | ||
| Gene Symbol: Gm6251 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001101561.2 | NP_001095031.2 | 1 | 129 | ||
| GenBank mRNA | CN721608.1 | - | 1 | 129 | ||

Rpl32
Genomic Map