siRNA Details
| Gene Symbol: Celf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001110228.1 | NP_001103698.1 | 10 | 1143 | ||
| NM_001110229.1 | NP_001103699.1 | 11 | 1230 | |||
| NM_001110230.1 | NP_001103700.1 | 11 | 1230 | |||
| NM_001110231.1 | NP_001103701.1 | 13 | 1205 | |||
| NM_001110232.1 | NP_001103702.1 | 12 | 1216 | |||
| NM_001160292.1 | NP_001153764.1 | 11 | 1230 | |||
| NM_001160293.1 | NP_001153765.1 | 10 | 1143 | |||
| NM_010160.2 | NP_034290.2 | 10 | 1143 | |||
| GenBank mRNA | AF090696.1 | - | 12 | 1216 | ||
| AF090697.1 | - | 10 | 1116 | |||
| AK137292.1 | - | 11 | 1171 | |||
| AK151818.1 | - | 9 | 1001 | |||
| AK152628.1 | - | 9 | 1001 | |||
| AK153267.1 | - | 9 | 1001 | |||
| AK159996.1 | - | 11 | 1235 | |||
| AK160861.1 | - | 13 | 1206 | |||
| AK161349.1 | - | 3 | 539 | |||
| AK164914.1 | - | 11 | 1171 | |||
| BC026856.1 | - | 10 | 1094 | 4503295 | ||
| Y18298.1 | - | 9 | 1009 | |||

Celf2
Genomic Map