siRNA Details
| Gene Symbol: Rhox2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_029203.2 | NP_083479.1 | 3 | 530 | ||
| GenBank mRNA | AK015997.1 | - | 3 | 531 | ||
| BC132216.1 | - | 3 | 524 | 40130493 | ||
| BC132220.1 | - | 3 | 524 | 40130497 | ||
| BC145534.1 | - | 3 | 524 | |||
| DQ058639.1 | - | 3 | 531 | |||
| Gene Symbol: Rhox2b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099316.1 | NP_001092786.1 | 3 | 508 | ||
| GenBank mRNA | BC049546.1 | - | 508 | |||
| Gene Symbol: Rhox2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099318.1 | NP_001092788.1 | 530 | |||
| Gene Symbol: Rhox2e | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001085348.1 | NP_001078817.1 | 530 | |||

Rhox2a
Genomic Map