siRNA Details
Gene Symbol: Coq10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081040.1 | NP_001074509.1 | 1, 2 | 1338 | ||
GenBank mRNA | BC147716.1 | - | 1, 2 | 487 | ||
BC147722.1 | - | 1, 2 | 487 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Coq10a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081040.1 | NP_001074509.1 | 1, 2 | 1338 | ||
GenBank mRNA | BC147716.1 | - | 1, 2 | 487 | ||
BC147722.1 | - | 1, 2 | 487 |