siRNA Details
Gene Symbol: Fam227b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029455.3 | NP_083731.2 | 4 | 267 | ||
GenBank mRNA | AK015893.1 | - | 4 | 249 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Fam227b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029455.3 | NP_083731.2 | 4 | 267 | ||
GenBank mRNA | AK015893.1 | - | 4 | 249 |